Anti-PSMB9 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8205-0025 PW8205-0100
89162-060EA 213.64 USD
89162-060 89162-062
Anti-PSMB9 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat proteasome 20S (beta1i subunit).

Type: Primary
Antigen: PSMB9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR