Anti-PSMB2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8145-0025 PW8145-0100
89162-016EA 178.76 USD
89162-016 89162-018
Anti-PSMB2 Mouse Monoclonal Antibody
Antibodies
Various systems for the nomenclature of the proteasome subunits have been established. This may be a source of confusion as the system on UniProt differs from "standard" nomenclature as described in the literature. The UniProt ID and Gene Name will help to clearly identify the proteins.

  • For Immunohistochemistry, Western Blot

Recognizes human and yeast proteasome 20S (beta2 subunit).

Type: Primary
Antigen: PSMB2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR