Anti-PSMA3 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8115-0025 PW8115-0100
89161-992EA 164.59 USD
89161-992 89161-994
Anti-PSMA3 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and rabbit proteasome 20S (alpha3 subunit).

Type: Primary
Antigen: PSMA3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR