Anti-PSMA6 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
BML-PW8100-0025 BML-PW8100-0100
89161-980EA 178.76 USD
89161-980 89161-982
Anti-PSMA6 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human and rabbit proteasome 20S (alpha6 subunit). Does not cross-react with rat or mouse protein.

Type: Primary
Antigen: PSMA6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR