Anti-UBA6 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0525-0100 PW0525-0025
89161-890EA 525 USD
89161-890 89161-888
Anti-UBA6 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human UBA6.

Type: Primary
Antigen: UBA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR