Cerulenin ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G237-0005 BML-G237-0025
89158-596EA 163.5 USD
89158-596 89158-598
Cerulenin ≥98% (by TLC)
Cerulenin

Fatty acid synthase inhibitor


Antibiotic. Fatty acid synthase (FAS) inhibitor reported to bind in equimolar ratio to β-keto-acyl-ACP synthase (KAS; IC50=1.5µM), thus inhibiting protein acylation at concentrations of 45-134 µM. Produces metabolic effects similar to the effects of leptin, but through mechanisms that are independent of, or down-stream from, both leptin and melanocortin receptors. Apoptosis inducer. Inhibitor of bacterial fatty acid synthesis (inhibits FabH, FabB and FabF condensation enzymes).

Formula: C₁₂H₁₇NO₃
MDL Number: MFCD00077686
CAS Number: 17397-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c873840fa907249383d2aa2ce069354cf3bb4d3&vid=15ad14f72d72dae64fe6b7ef89a6d0435546634c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR