Furoxan ≥99% (by HPLC)

Supplier: Enzo Life Sciences

BML-CN255-0010 BML-CN255-0050
89157-062EA 95.92 USD
89157-062 89157-064
Furoxan ≥99% (by HPLC)
Furoxan

A novel NO donor which only releases NO in the presence of thiols. It is a potent vasodilator and inhibitor of platelet aggregation.


  • Releases NO in the presence of thiols


White solid.


Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml).

Formula: C₉H₅N₃O₂
Melting Pt: 72…74 °C
Storage Temperature: Freezer
CAS Number: 125520-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05e55818bd31a92ea2de1fc15e47ddad2165dd1d&vid=ab97e42541b4aa85e67daa78f9d929992923e1b1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR