SCREEN-WELL® Phosphatase Inhibitor Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2834-0100
BML-2834-0500
89156-298EA
1660.07
USD
89156-298
89156-300
SCREEN-WELL® Phosphatase Inhibitor Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Screen-Well™ Phosphatase Inhibitor Library contains 33 known phosphatase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 µl or 500 µl per well.
The library is ideal for chemical genomics, assay development, and as a reference set for secondary screening. Includes inhibitors of these important phosphatases: Calcineurin(PP2B), JSP-1, PRL-1, CD45, PP1, PRL-3, CDC25, PP2A, PTEN, and many more.