Anti-TRIP-BR1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-645-C050 804-645-C100
89154-694EA 367.33 USD
89154-694 89154-696
Anti-TRIP-BR1 Mouse Monoclonal Antibody
Antibodies
TRIP-Br1 (SEI-1) functions as physiological integrator of transcriptional regulatory signals at E2F-responsive promoters to ensure proper execution of E2F-dependent cell cycle progression. TRIP-Br1 is defined as an oncogene.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, pig and monkey TRIP-Br1.

Type: Primary
Antigen: TRIP-BR1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR