Anti-CD40L Hamster Monoclonal Antibody

Supplier: Enzo Life Sciences
804-331-C100 804-331-C500
89154-052EA 391.31 USD
89154-052 89154-054
Anti-CD40L Hamster Monoclonal Antibody
Antibodies
Host: Hamster, Isotype: IgG1

Recognizes mouse CD40L.

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR