(+)-Bicuculline ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

550-515-M050 550-515-M250
89152-718EA 73.03 USD
89152-718 89152-720
(+)-Bicuculline ≥98% (by HPLC)
(+)-Bicuculline

GABAA receptor antagonist.


  • Formulation: Off-white to yellow powder.


Soluble in benzene, chloroform or ethyl acetate.

Formula: C₂₀H₁₇NO₆
Storage Temperature: Freezer
MDL Number: MFCD00005006
CAS Number: 485-49-4
EINECS: 207-619-7
UN: 1544
ADR: 6.1,II
Merck Index: 14,01203
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2643bc2d0e6d659e85983cb6e28215dbef6184d8&vid=250347e0a96d93d4ce0249beb1e2e0bb27bba1d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Appearance Off-white to yellow powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR