Tamoxifen citrate ≥99%

Supplier: Enzo Life Sciences

Synonyms: (Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate

550-095-G001
89152-148EA 157 USD
89152-148
Tamoxifen citrate ≥99%
Tamoxifen citrate

PKC inhibitor


Antiestrogen with anti-cancer activity. Selective protein kinase C (PKC) inhibitor and induces PKC epsilon translocation. Induces apoptosis in estrogen receptor-negative human cancer cell lines.

Formula: C₂₆H₂₉NO
MW: 563.65 g/mol
MDL Number: MFCD00058321
CAS Number: 54965-24-1
https://us.vwr.com/stibo/web/std.lang.all/08/06/45390806.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR