Me-7BIO ≥98%

Supplier: Enzo Life Sciences
Warning

Synonyms: (2’Z,3’E)-7-Bromo-1-methylindirubin-3’-oxime

430-157-M001
89151-274EA 152 USD
89151-274
Me-7BIO ≥98%
Me-7BIO

Control compound for 6BIO.


Soluble in DMSO.

https://us.vwr.com/stibo/web/std.lang.all/07/04/45390704.jpg

Order Now

Specification Test Results

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR