Lupeol ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
Fagarasterol, Monogynol B, Clerodol, 20(29)-Lupen-3β-ol, 3β-Hydroxy-20(29)-lupene
350-355-M010
350-355-M050
89149-572EA
1730
USD
89149-572
89149-574
89149-576
Lupeol ≥98% (by HPLC)
Lupeol
Anti-inflammatory, antioxidant and antimutagenic compound
Anti-inflammatory, antioxidant and antimutagenic compound. Shows preventive effects on DMBA induced DNA alkylation damage in mouse skin. Modulator of NF-κB and PI(3)K/Akt (protein kinase B; PKB) pathways. Induces Fas-mediated apoptosis via inhibition of Ras signalling. Inhibits tumor growth in mouse in vivo.
Isolated from Lupinus polyphyllus.
Formula:
C₃₀H₅₀O MW: 426.7 g/mol |
CAS Number:
545-47-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...