Pseudohypericin ≥98% (by HPLC, TLC)
Supplier: Enzo Life Sciences
350-276-M001 350-276-M005
89149-392EA 175.49 USD
89149-392 89149-394
Pseudohypericin ≥98% (by HPLC, TLC)
Pseudohypericin
Inhibitor of PKC and dopamine beta-hydroxylase
Inhibitor of protein kinase C (PKC) (IC50=15µg/ml). Antiviral and antiretroviral activity. Potent inhibitor of dopamine-beta-hydroxylase (IC50=3µg/ml).
Isolated from Hypericum perforatum.
CAS Number: 55954-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fa4824321441cddf01249694ec65742306e0c49&vid=4a5368aeae1554bbc19c29318d32ac1b74f6738d&prid=web&clid=SAPDAM