myo-Inositol ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Inositolrel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaoli-InositolD-Myo-Inositol

307-003-G025
89148-752EA 50.2 USD
89148-752
myo-Inositol ≥98% (by HPLC)
myo-Inositol

  • Formulation: White to off-white solid.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
MDL Number: MFCD00077932
CAS Number: 87-89-8
https://us.vwr.com/stibo/web/std.lang.all/76/33/45347633.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR