N-Acetyl-L-cysteine ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Acetylcysteine(R)-2-Acetamido-3-mercaptopropanoic acid(R)-(+)-N-AcetylcysteineAc-Cys-OH

105-005-G005
89142-808EA 51.3 USD
89142-808
N-Acetyl-L-cysteine ≥98% (by TLC)
N-Acetyl-L-cysteine

Derivative of cysteine wherein an acetyl group is attached to the N. Is used as a specific antidote in cases of acetaminophen overdose.


  • Formulation: White powder.

Formula: C₅H₉NO₃S
MW: 163.2 g/mol
MDL Number: MFCD00004880
CAS Number: 616-91-1
https://us.vwr.com/stibo/web/std.lang.all/74/18/45347418.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR