Anti-GRP75 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPS-825-D ADI-SPS-825-F
200062-834EA 417.47 USD
200062-834 95043-774
Anti-GRP75 Mouse Monoclonal Antibody
Antibodies
Grp75 (Glucose regulated 75 kDa protein) is a member of the Hsp70 family. It is localized in the mitochondrial matrix, where, in concert with Hsp60, is thought to participate in the re-folding of proteins translocated into this organelle. Like its E. coli homolog DnaK, Grp75 possesses a cation-dependent ATPase activity thought to be central to its function as a chaperone.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, chicken, Drosophila, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Grp75. Detects a band of ~75kDa by Western blot.

Type: Primary
Antigen: GRP75
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR