Anti-HSPA8 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-819-D ADI-SPA-819-F
95043-808EA 222.36 USD
95043-808 95043-810
Anti-HSPA8 Rabbit Polyclonal Antibody
Antibodies
The 70 kDa heat shock protein Hsp70 belongs to the Hsp70 family of highly-related protein isoforms ranging in size from 66 kDa to 78 kDa. Hsc70 shares close biochemical and biological ties to Hsp70, and also belongs to the Hsp70 family. These proteins include cognate members found within major intracellular compartments and highly inducible isoforms predominantly cytoplasmic or nuclear in distribution. Members of the Hsp70 family function as molecular chaperones involved in such cellular functions as protein folding, transport, maturation and degradation, operating in an ATP-dependent manner. The molecular chaperones of the Hsp70 family recognize and bind to nascent polypeptide chains or partially folded intermediates of proteins, preventing their aggregation and misfolding, and the binding of ATP triggers a critical conformational change leading to the release of the bound substrate protein. Data demonstrates that with a ubiquitin-like domain at its amino terminus and its association with the 26S proteosome in HeLa cells, Bag-1 modulates the chaperone activity of Hsc70 and Hsp70. These findings reveal Bag-1's role as a physical link between the Hsc70/Hsp70 chaperone system and the proteasome. Experimental data also shows that the ATPase domain and the substrate binding domain of Hsd70 cooperate to form a co-chaperone-chaperone complex with the synaptic vesicle cysteine string protein (csp), essential for normal neurotransmitter release.

  • For Western Blot

Recognizes human, mouse and rat HSC70 (HSP73). Detects a band of ~73kDa by Western blot.

Type: Primary
Antigen: HSPA8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR