Stop Solution 2, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-80-1804 ADI-80-0377
95045-200EA 91.56 USD
95045-200 95052-788
Stop Solution 2, Enzo Life Sciences
Assays ELISAs
Acid stop solution (1 N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions.
Each 50 ml bottle is sufficient for use with 5×96 wells microtiter plates.
Ordering information: Additional assay development reagents are available as companion products to IMMUNOSET® products.