Mouse;Rat beta-Amyloid (1-42)
Supplier: BACHEM AMERICAS INC
4035885.0500 4035885.1000
H-5966.0500BAEA 638.85 USD
H-5966.0500BA H-5966.1000BA
Mouse;Rat beta-Amyloid (1-42)
Proteins and Peptides
Rodent Aβ 1-42 is considerably less aggregation-prone than human Aβ 1-42, but antimicrobial activity against a range of clinically relevant microorganisms has been shown for both peptides.