(D-Ala¹)-Peptide T amide
Supplier: BACHEM AMERICAS INC
4015906.0005 4015906.0025
H-5935.0005BAEA 392.15 USD
H-5935.0005BA H-5935.0025BA
(D-Ala¹)-Peptide T amide
Proteins and Peptides
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
Potent analog of peptide T. DAPTA (aSTTTNYT-amide) showed a strong anti-HIV 1 activity in monocytes/macrophages, the peptide inhibits the entry of the virus.