Mouse;Rat PAR-2 (1-6) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4026881.0005 4026881.0025
H-3586.0005BAEA 326.8 USD
H-3586.0005BA H-3586.0025BA
Mouse;Rat PAR-2 (1-6) Trifluoroacetate
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
Tethered ligand sequence of mouse PAR-2, which like the thrombin receptor is a protease-activated receptor present in keratinocytes. SLIGRL was found to be equipotent with SFLLRN (H-8365) in activating keratinocyte inositolphospholipid hydrolysis and calcium mobilization.