Dog;Human;Pig GRP (18-27)
Supplier: Bachem
4011670.0005 4011670.0001
H-3120.0001BAEA 128.72 USD
H-3120.0001BA H-3120.0005BA
Dog;Human;Pig GRP (18-27)
Proteins and Peptides
Bombesin-like peptide from porcine spinal cord; exhibits a potent stimulant effect on smooth muscle of rat uterus. Neuromedin C microinjected into the amygdala inhibited feeding in rats.