(D-Ala²,D-Leu⁵)-Enkephalin
Supplier: BACHEM AMERICAS INC
4001508.0025 4001508.0100
H-2860.0025BAEA 251.47 USD
H-2860.0025BA H-2860.0100BA
(D-Ala²,D-Leu⁵)-Enkephalin
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
DADLE has generally served as the putative, prototypical δ-opioid receptor ligand. DADLE is more potent and selective than Leu-enkephalin or Met-enkephalin in the MVD versus GPI bioassay systems and more potent and approximately as selective as Leu-enkephalin in binding assays. It shows roughly the same antinociceptive activity as β-endorphin.