Angiotensin I Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4002346.0025 4002346.0100
H-1680.0025BAEA 206.12 USD
H-1680.0025BA H-1680.0100BA
Angiotensin I Trifluoroacetate
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
Angiotensin I, DRVYIHPFHL, is cleaved by angiotensin-converting enzyme (ACE) yielding Ang II (H-1705) and His-Leu (G-2310). Ang I is also converted to Ang II by chymase, especially in the human heart. Hence, this mast cell protease is also involved in blood pressure regulation. The chymase pathway gains importance in inflammatory conditions.