Albumin, Human, RIA Grade, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
08823030
08823032
IC823030GM
149.7
USD
IC823030
IC823032
Albumin, Human, RIA Grade, MP Biomedicals
Proteins and Peptides
1X Crystalline Lyophilized. Purity is 97 to 99%. Low B12 and Folate. Prepared using Cohn method IV. Source material is non-reactive and is tested for Hbs Ag, Anti-HCV, Anti-HIV-1, Anti-HIV-2, and Syphillis.
- Moisture: ≤5%