ImmunO™ Bovine Serum Albumin (BSA)

Supplier: MP Biomedicals
ImmunO™
08810061 08810063
IC810061EA 483.15 USD
IC810061 IC810063
ImmunO™ Bovine Serum Albumin (BSA)
Proteins and Peptides
Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.

  • Content: Sodium Chloride: 0.8 g/dL
  • Osmolality: Reported
  • Sodium: 0.8 g/dL
  • Specific gravity: Reported
  • Refractive Index: Reported
  • Optical rotation: [a]259: -61°; [a]264: -63° (Lit.)
  • Isoelectric point (pl): In water at 25 °C: Endogenous Material - 4.7; 4.9; Fatty Acid Depleted - 5.3(Lit.)
  • Sterile
  • Material prepared from bovine albumin fractionated and purified from bovine plasma or serum
  • Collected from USDA approved facilities

Albumin is used to solubilize lipids, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR