ImmunO™ Bovine Serum Albumin (BSA), MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

08810661 08810667
IC810661EA 220.85 USD
IC810661 IC810667
ImmunO™ Bovine Serum Albumin (BSA), MP Biomedicals
Proteins and Peptides

Albumin bovine is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates.


  • Content: Calcium 0.02 mg/g; Total Lipids 0.2 mg/g
  • Reagent Grade
  • Purity 100.0%
  • Heat-shocked, IgG-free, low free fatty acids
  • Heavy metals: 4.7pm
  • Loss on Drying: <5.0%
  • Optical rotation: [a]259: -61°; [a]264: -63° (Lit.)


Albumin is used to solubilize lipids, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).


At pH 5-7 it contains 17 intrachain disulfide bridges and 1 sulfhydryl group.


Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR