ImmunO™ Bovine Serum Albumin (BSA), MP Biomedicals

Supplier: 0000012207

08810032 08810033 08810034
IC810032EA 404.63 USD
IC810032 IC810033 IC810034
ImmunO™ Bovine Serum Albumin (BSA), MP Biomedicals
Proteins and Peptides

Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.


  • Proteases: <5 ng/mg
  • Sodium: ≤10 mg/g
  • Chloride: ≤3 mg/g
  • Heavy metals:<10 ppm
  • Iron: ≤5 mg/g
  • Loss on drying: 0.007
  • Conductivity :<3000 mS (10%)
  • Negative for Bluetongue and Vesicular Stomatitis Virus
  • IgG, Protease-free
  • Optical rotation: [a]259: -61°; [a]264: -63° (Lit.)
  • Absorbance: <0.1 (4%) @ 440nm


Albumin is used to solubilize lipids, and is also used as a blocking agent in Western blots or ELISA applications. Globulin free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).


Prior to lyophilization, the product is filtered through a 0.2 µm filter.


Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR