Gelatin

Supplier: Thermo Scientific Chemicals

J62699-EQE J62699-AK
AAJ62699-EQEEA 134.8 USD
AAJ62699-EQE AAJ62699-AK
Gelatin
Proteins and Peptides

Liquid


CAS No.: 9000-70-8
Physical Form: Liquid
MDL No.: MFCD00081638

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR