DNA (Lambda Phage) Hind III Digest Molecular Weight Marker, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-105-0100
RLMB-105-0100EA 131.64 USD
RLMB-105-0100
DNA (Lambda Phage) Hind III Digest Molecular Weight Marker, Rockland Immunochemicals
Electrophoresis Reagents Molecular Weight Markers
Lambda DNA fragments prepared by the digestion of purified lambda DNA with the restriction endonuclease Hind III
On agarose gel electrophoresis the mixture separates into eight individual bands having the following number of base pairs: 23130, 9416, 6557, 4361, 2322, 2027, 564, and 125.