Anti-IgG Sheep Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
612-6602
RL612-6602EA 177.61 USD
RL612-6602
Anti-IgG Sheep Polyclonal Antibody (Biotin)
Antibodies
This product has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Sheep
Isotype: IgG
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen Symbol IgG
Environmentally Preferable
Antigen Name Immunoglobulin G
Antibody Type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Rat
Host Sheep
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope Heavy and Light chain
Immunogen Rat IgG whole molecule
Format Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Concentration 2.0 mg/ml by UV absorbance at 280 nm
Cross Adsorption No
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage Temperature –20 °C
Shipping Temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR