Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
611-1624
RL611-1624EA 287.31 USD
RL611-1624
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
This product has been assayed against 1.0 µg of Rabbit IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

SPECIFICATIONS

Antigen Symbol IgG
Environmentally Preferable
Antigen Name Immunoglobulin G
Antibody Type Secondary
Clonality Polyclonal
Conjugation Biotin
Reactivity Rabbit
Host Goat
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Epitope F(ab')2
Immunogen Rabbit IgG F(ab')2 fragment
Format Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Concentration 2.0 mg/ml by UV absorbance at 280 nm
Cross Adsorption No
Storage Buffer 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin. Immunoglobulin and Protease free.
Storage Temperature –20 °C
Shipping Temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR