Anti-HIST4H4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-J01
RL600401J01EA 575.67 USD
RL600401J01
Anti-HIST4H4 Rabbit Polyclonal Antibody
Antibodies
Anti-Histone H4 [Monomethyl Lys20] antibody is useful for Western Blot, Dot Blot, Chromatin Immunoprecipitation, Immunofluorescence, and Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band approximate

Recommended Dilutions: ChIP: 2-5µg/million cells; Immunofluorescence: 1:50; Western Blot: 1:500; Immunochemistry: 1:50; Other Dilution: Dot Blot 1:1000

Type: Primary
Antigen: HIST4H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, C. elegans, Rat, Chicken, Xenopus, Drosophila, plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR