Anti-VDAC1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-882S
RL600401882SEA 151.49 USD
RL600401882S
Anti-VDAC1 Rabbit Polyclonal Antibody
Antibodies
VDAC/Porin Antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~30-33 kDa in size.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:500 - 1:2000; Immunochemistry: Yes; Other Dilution: User Optimized

Type: Primary
Antigen: VDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human, Rabbit, Bovine, Pig, Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR