Anti-CALCA Mouse Monoclonal Antibody [clone: 11D9.F3.C12.B8]

Supplier: Rockland Immunochemical
200-301-I44
RL200-301-I44EA 565.22 USD
RL200-301-I44
Anti-CALCA Mouse Monoclonal Antibody [clone: 11D9.F3.C12.B8]
Antibodies
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 11D9.F3.C12.B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR