Anti-CALCA Mouse Monoclonal Antibody [clone: 18C4.B4.C2]

Supplier: Rockland Immunochemical
200-301-I40S
RL200-301-I40SEA 151.49 USD
RL200-301-I40S
Anti-CALCA Mouse Monoclonal Antibody [clone: 18C4.B4.C2]
Antibodies
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 18C4.B4.C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR