Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL1] (FITC)
Supplier: STEMCELL Technologies
60097FI
MSPP-60097FIEA 374.15 USD
MSPP-60097FI
Anti-CD45RO Mouse Monoclonal Antibody [clone: UCHL1] (FITC)
Antibodies
Mouse monoclonal IgG2a antibody against human, chimpanzee, common marmoset CD45RO, FITC-conjugated.
  • Leukocytes
  • T cells
  • T cells CD4
  • T cells CD8
The UCHL1 antibody reacts with an extracellular epitope on CD45RO, which is the shortest isoform of CD45, a type I transmembrane glycoprotein and member of the protein tyrosine phosphatase family (receptor class 1/6 subfamily). Alternative splicing of exons 4, 5 and 6 that encode the extracellular RA, RB and RC polypeptides of CD45 gives rise to up to 8 isoforms with molecular masses of 180 - 240 kDa. ExcisionMore Product Information