Anti-TRA-2-49 Mouse Monoclonal Antibody [clone: TRA-2-49/6E]
Supplier: STEMCELL Technologies
60066
MSPP-60066EA 840.38 USD
MSPP-60066
Anti-TRA-2-49 Mouse Monoclonal Antibody [clone: TRA-2-49/6E]
Antibodies
Mouse monoclonal IgG1 antibody against human, chimpanzee, gibbon TRA-2-49 (tissue non-specific alkaline phosphatase), unconjugated.
  • Pluripotent stem cells
The TRA-2-49/6E antibody reacts with the tissue non-specific isoform of alkaline phosphatase (AP), which is expressed at high levels in undifferentiated pluripotent stem cells such as human and mouse embryonic stem (ES) cells, as well as induced pluripotent stem (iPS) and embryonal carcinoma (EC) cells. This isozyme of AP is the same as that expressed by liver, bone and kidney cells. Levels of APMore Product Information