Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2] (Alexa Fluor® 488)
Supplier: STEMCELL Technologies
60036AD 60036AD.1
MSPP-60036ADEA 640.15 USD
MSPP-60036AD MSPP-60036AD1
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2] (Alexa Fluor® 488)
Antibodies
Rat monoclonal IgG2a antibody against mouse CD150 (SLAM), Alexa Fluor® 488-conjugated.
  • Hematopoietic Stem
  • Progenitor cells
The TC15-12F12.2 antibody reacts with CD150 (signaling lymphocyte activation molecule or SLAM), an ~75 kDa type I transmembrane glycoprotein that plays multiple roles in the immune response by serving as a cell adhesion molecule and/or coreceptor. It is differentially expressed by T cells, immature thymocytes, B cells, dendritic cells, macrophages, and endothelial cells. The expression pattern differs according to cell typeMore Product Information