EasySep™ Mouse Plasmacytoid DC Isolation Kits
Supplier: STEMCELL Technologies
19764 19764RF
MSPP-19764EA 1389.23 USD
MSPP-19764 MSPP-19764RF
EasySep™ Mouse Plasmacytoid DC Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched mouse plasmacytoid dendritic cells (pDC)
  • Fast, easy-to-use and column-free
  • Up to 94% purity
  • Isolated cells are untouched
The EasySep™ Mouse Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-pDCs. Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotinMore Product Information