EasySep™ Human CD4+ T Cell Isolation Kits
Supplier: STEMCELL Technologies
17952 17952RF 100-0696
MSPP-17952EA 1483.08 USD
MSPP-17952 MSPP-17952RF MSPP-100-0696
EasySep™ Human CD4+ T Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human CD4+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 97% purity with high recovery
  • Untouched, viable cells
The EasySep™ Human CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells byMore Product Information