EasySep™ Human Central and Effector Memory CD8+ T Cell Isolation Kit
Supplier: STEMCELL Technologies
17869
MSPP-17869EA 1533.85 USD
MSPP-17869
EasySep™ Human Central and Effector Memory CD8+ T Cell Isolation Kit
Cell Separation Media
Immunomagnetic positive selection of human central and effector memory CD8+ T cells.
  • Easy-to-use and column-free
  • Up to 93% purity
  • No-wash removal of EasySep™ Releasable RapidSpheres™
This kit isolates human CD62L+CD45RO+ and CD62L-CD45RO+ memory CD8+ T cells from fresh peripheral blood mononuclear cells (PBMCs) or washed leukapheresis samples. First, CD62L+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the EasySep™-More Product Information