Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2] (APC)
Supplier: STEMCELL Technologies
100-1453
MSPP-100-1453EA 356.62 USD
MSPP-100-1453
Anti-IL-4 Rat Monoclonal Antibody [clone: MP4-25D2] (APC)
Antibodies
Rat monoclonal antibody against human IL-4, APC-conjugated.
  • T cells CD4
This rat monoclonal antibody (clone MP4-25D2) reacts with human interleukin 4 (IL-4). IL-4 is an ~15 kDa protein primarily composed of a four-helix bundle (Powers et al.) and is produced by a host of immune cells including T helper 2 (Th2) cells, basophils, eosinophils, and mast cells (Gadani et al.). It exhibits high affinity for the IL-4 receptor alpha (IL-4Rα) chain, which dimerizes with either the common gamma chain (γc) or IL-13 receptor alpha 1 (IL-13Rα1), leading toMore Product Information