HepatiCult™ Organoid Kit (Human)
Supplier: STEMCELL Technologies
100-0386
MSPP-100-0386EA 1941.54 USD
MSPP-100-0386
HepatiCult™ Organoid Kit (Human)
Cell Culture Media
Culture medium kit for initiation, growth, and differentiation of human liver organoids.
Reliably generate, expand, and differentiate physiologically relevant human liver organoids. Compared to traditionally used model systems such as cell lines and animal models, human liver organoids provide a more relevant model system for studying hepatic development, regeneration, metabolism, and disease.
HepatiCult™ Organoid Kit (Human) contains all the necessary components to culture human liver organoids from fresh or cryopreserved human liver tissue:
HepatiCult™ Organoid Initiation Medium (Human) for efficient generation of hepatic organoids across different donor lines
HepatiCult™ Organoid Growth Medium (Human) for expansion and maintainence of hepatic organoids for future experimentation or bio-banking
HepatiCult™ Organoid Differentiation Medium (Human) for generating mature organoids that demonstrate hepatic functionality, including CYP3A4 activity.
These organoids can be adapted to a range of culture protocols, including 2D monolayer, suspension cultures, and high-throughput assays For more information on protocols for organoid culture with HepatiCult™, please explore the technical manual and educational materials. Should you intend to use this product for commercial.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').