STEMdiff™ Atrial Cardiomyocyte Differentiation Kit
Supplier: STEMCELL Technologies
100-0215
MSPP-100-0215EA 1324.62 USD
MSPP-100-0215
STEMdiff™ Atrial Cardiomyocyte Differentiation Kit
Cell Culture Media
Serum-free culture medium kit for differentiation of human PSCs to atrial cardiomyocytes.
STEMdiff™ atrial cardiomyocyte differentiation kit is optimized for the differentiation and maintenance of human embryonic stem and induced pluripotent stem cells into atrial cardiomyocytes (cardiac troponin T-positive; cTnT+). This serum-free kit can be used to generate atrial cardiomyocytes derived from a clump culture of human pluripotent stem cells (hPSCs) maintained in mTeSR™1, mTeSR™ plus, or TeSR™-E8™. With greater than 80% yield purity, an average of 1×10⁶ cTnT+ cells can be harvested from a single well of a 12-well plate. STEMdiff™ cardiomyocyte maintenance kit comprises the maintenance basal medium and supplement and allows for long-term maintenance of hPSC-derived cardiomyocytes for one month or longer. These cardiomyocytes can be used in various downstream applications and analyses.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').