STEMdiff™ Mesoderm Induction Medium
Supplier: STEMCELL Technologies
05221
MSPP-05221EA 675.38 USD
MSPP-05221
STEMdiff™ Mesoderm Induction Medium
Cell Culture Media
Defined, xeno-free induction medium for early mesodermal differentiation.
STEMdiff™ Mesoderm Induction Medium (MIM) is a defined, xeno-free medium for generation of early mesoderm cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells. Protocols for mesodermal differentiation can be difficult and inconsistent, therefore, use the short and simple STEMdiff™ MIM monolayer protocol to differentiate your human pluripotent stem cells (hPSCs). STEMdiff™ MIM is a complete medium that produces a cell population enriched for early mesoderm, as indicated by positive expression of Brachyury (T) and NCAM markers. As part of the hPSC workflow, STEMdiff™ MIM efficiently differentiates hPSCs cultured in TeSR™ media. When directed, early mesoderm cells produced using STEMdiff™ MIM can be further differentiated to specialized cell types, such as osteoblasts, chondrocytes, adipocytes or endothelial cells.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').