Human IL-1 beta ELISA Kit
Supplier: STEMCELL Technologies
02004
MSPP-02004EA 1460.08 USD
MSPP-02004
Human IL-1 beta ELISA Kit
Assays ELISAs
For detection and measurement of human interleukin 1 beta.
The Human Interleukin 1 beta (IL-1β) ELISA Kit is designed for the quantitative detection and measurement of human IL-1β in biological fluids such as serum, plasma, and cell culture supernatants. IL-1β is secreted by monocytes, macrophages, and dendritic cells. IL-1β is a proinflammatory cytokine that recruits immunocompetent cells to affected sites by stimulating the expression of adhesion molecules.
The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine. The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample. The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').