QuikChange II, Agilent Technologies
Supplier: AGILENT TECH
200523 200524 200555 200521 200522
99900-068EA 389.48 USD
99900-068 99900-074 99900-098 ST200521 ST200522
QuikChange II, Agilent Technologies
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
These QuikChange II kits provide improved fidelity, while maintaining greater than 80% mutation efficiency for single site mutagenesis.
  • Includes PfuUltra High-fidelity DNA polymerase to minimize unwanted errors
  • Complete kit provides all reagents necessary to support mutagenesis (at single sites)
Used for large constructs, and for use with electroporation competent cells.
Caution: For research use only. Not for use in diagnostic procedures.